Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Chloro-11-[(n-methyl)-4-piperidylidene]-6,11-dihydro-5h-benzo [5,6] cyclohepta[1,2b]pyridine 97%

Supplier: AMBEED, INC
Warning

A539310-1G A539310-5G A539310-100G A539310-100MG A539310-250MG A539310-25G A539310-1G
103828-618EA 9 USD InStock
103828-618 103839-740 76843-786 76980-170 76980-172 76980-174 76980-176 77307-766
8-Chloro-11-[(n-methyl)-4-piperidylidene]-6,11-dihydro-5h-benzo [5,6] cyclohepta[1,2b]pyridine 97%
8-Chloro-11-[(n-methyl)-4-piperidylidene]-6,11-dihydro-5h-benzo [5,6] cyclohepta[1,2b]pyridine
Precaución: For research use only


Formula: C₂₀H₂₁ClN₂
MW: 324.847 g/mol
Boiling Pt: 474.3 °C (760 mm Hg)
Melting Pt: 106…109 °C
Flash Pt: 240.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06411084
CAS Number: 38092-89-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty