Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Hydroxyquinoline-2-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A130916-1G A130916-10G A130916-250MG A130916-1G A130916-25G A130916-5G
103801-490EA 40 USD InStock
103801-490 76797-852 76797-854 77313-962 77313-964 77313-966
8-Hydroxyquinoline-2-carbaldehyde 98%
8-Hydroxyquinoline-2-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
Melting Pt: 97…100 °C
Flash Pt: 191 °C
Storage Temperature: Ambient
MDL Number: MFCD00168962
CAS Number: 14510-06-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty