Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole 97%

Supplier: AMBEED, INC
Warning

A263708-1G A263708-5G A263708-250MG A263708-5G A263708-25G
103812-638EA 7 USD InStock
103812-638 103852-528 77238-584 77304-878 77746-451
9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole 97%
9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
Precaución: for Research Use Only


Formula: C₂₄H₂₄BNO₂
Boiling Pt: 483.2±37.0 °C (760 mm Hg)
Flash Pt: 246 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22415352
CAS Number: 870119-58-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty