Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(9-Carbazolyl)benzeneboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A686553-5G A686553-1G A686553-25G
103840-560EA 17 USD InStock
103840-560 76844-774 76844-776
4-(9-Carbazolyl)benzeneboronic acid pinacol ester 98%
4-(9-Carbazolyl)benzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₂₄H₂₄BNO₂
MW: 369.27 g/mol
Boiling Pt: 468.5 °C (760 mm Hg)
Melting Pt: 165…173 °C
Flash Pt: 237.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16294549
CAS Number: 785051-54-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty