Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-(4-Bromophenyl)-10-phenylanthracene 98%

Supplier: AMBEED, INC
Warning

A389647-1G A389647-5G A389647-100G A389647-10G A389647-250MG A389647-25G
103826-848EA 11 USD InStock
103826-848 103837-888 76841-250 76841-252 77016-166 77016-168 77016-170 77306-994
9-(4-Bromophenyl)-10-phenylanthracene 98%
9-(4-Bromophenyl)-10-phenylanthracene
Precaución: for Research Use Only


Formula: C₂₆H₁₇Br
Storage Temperature: Ambient
MDL Number: MFCD24842536
CAS Number: 625854-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty