Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anthracene-9,10-diboronic acid bis(pinacol) ester 98%

Supplier: AMBEED, INC
Warning

A306909-1G A306909-5G A306909-1G A306909-5G A306909-250MG
103814-486EA 32 USD InStock
103814-486 103835-658 76838-390 76838-392 77154-278
Anthracene-9,10-diboronic acid bis(pinacol) ester 98%
Anthracene-9,10-diboronic acid bis(pinacol) ester
Precaución: for Research Use Only


Formula: C₂₆H₃₂B₂O₄
Boiling Pt: 575.3 °C (760 mm Hg)
Flash Pt: 301.7 °C
Storage Temperature: Ambient
MDL Number: MFCD16294538
CAS Number: 863992-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty