Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Anthraceneboronic acid 98%

Supplier: AMBEED, INC
Warning

A112579-1G A112579-5G A112579-100G A112579-10G A112579-250MG A112579-25G A112579-5G A112579-1G
103798-044EA 8 USD InStock
103798-044 103823-882 77108-074 77108-076 77108-078 77108-080 77297-192 77310-692
9-Anthraceneboronic acid 98%
9-Anthraceneboronic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₁BO₂
MW: 222.05 g/mol
Melting Pt: 203…250 °C
Flash Pt: 243.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03425925
CAS Number: 100622-34-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty