Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Bromoanthracene 98%

Supplier: AMBEED, INC
Warning

A157372-5G A157372-100G A157372-10G A157372-500G A157372-25G A157372-5G
103843-198EA 5 USD InStock
103843-198 76783-682 76783-684 76783-686 77300-254 77319-016
9-Bromoanthracene 98%
9-Bromoanthracene
Precaución: for Research Use Only


Formula: C₁₄H₉Br
MW: 257.13 g/mol
Boiling Pt: 359
Melting Pt: 103
Flash Pt: 190.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty