Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Bromononanoic acid 98%

Supplier: AMBEED, INC
Warning

A296554-1G A296554-25G A296554-5G A296554-100G A296554-10G A296554-250MG A296554-1G
103814-104EA 14 USD InStock
103814-104 103835-246 76837-920 76837-922 77262-912 77262-914 77262-916 77305-696
9-Bromononanoic acid 98%
9-Bromononanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₇BrO₂
MW: 237.14 g/mol
Melting Pt: 39 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00046564
CAS Number: 41059-02-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty