Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxycarbazole 97%

Supplier: AMBEED, INC
Warning

A116407-5G A116407-100G A116407-10G A116407-25G A116407-5G
103824-594EA 6 USD InStock
103824-594 76718-316 76718-318 76718-320 77311-406
4-Hydroxycarbazole 97%
4-Hydroxycarbazole
Precaución: for Research Use Only


Formula: C₁₂H₉NO
Boiling Pt: 431.4 °C (760 mm Hg)
Melting Pt: 169…173 °C
Flash Pt: 214.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02178385
CAS Number: 52602-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty