Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9H-Pyrido[2,3-b]indole 95%

Supplier: AMBEED, INC
Warning

A247147-5G A247147-10G A247147-250MG A247147-1G A247147-25G A247147-5G
103811-750EA 9 USD InStock
103811-750 103851-582 76987-802 76987-804 77090-892 77090-894 77090-896
9H-Pyrido[2,3-b]indole 95%
9H-Pyrido[2,3-b]indole
Precaución: for Research Use Only


Formula: C₁₁H₈N₂
Boiling Pt: 372.9 °C (760 mm Hg)
Melting Pt: 210…212 °C
Flash Pt: 171.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00473709
CAS Number: 244-76-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty