Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3',4'-Dihydroxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A200746-1G A200746-5G A200746-250MG A200746-1G A200746-5G A200746-25G A200746-10G
103817-236EA 10 USD InStock
103817-236 103847-870 76691-170 77085-934 77085-936 77302-718 77600-638
3',4'-Dihydroxyacetophenone 98%
3',4'-Dihydroxyacetophenone
Precaución: for Research Use Only


Formula: C₈H₈O₃
Boiling Pt: 372 °C (760 mm Hg)
Melting Pt: 117 °C
Flash Pt: 193.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00016439
CAS Number: 1197-09-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty