Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amino(imino)methanesulfonic acid 95%

Supplier: AMBEED, INC
Warning

A131304-1G A131304-100MG A131304-250MG A131304-5G A131304-1G
103801-564EA 111 USD InStock
103801-564 77009-962 77009-964 77009-966 77298-472
Amino(imino)methanesulfonic acid 95%
Amino(imino)methanesulfonic acid
Precaución: for Research Use Only


Formula: CH₄N₂O₃S
Melting Pt: 132…134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19203404
CAS Number: 1184-90-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty