Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Apiin 98%

Supplier: AMBEED, INC
Warning

A101993-5MG A101993-25MG A101993-1MG A101993-50MG A101993-1MG A101993-5MG
103791-412EA 62 USD InStock
103791-412 103791-964 103794-990 103799-258 77282-882 77282-884
Apiin 98%
Apiin
Precaución: for Research Use Only


Formula: C₆H₁₀N₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00016941
CAS Number: 26544-34-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty