Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Azetidinecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A514053-5G A514053-100G A514053-500G A514053-1G A514053-25G
103828-434EA 9 USD InStock
103828-434 103839-536 76830-312 76830-314 76843-518 76843-520
3-Azetidinecarboxylic acid 97%
3-Azetidinecarboxylic acid
Precaución: for Research Use Only


Formula: C₄H₇NO₂
Melting Pt: 286 °C
Flash Pt: 100.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191763
CAS Number: 36476-78-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty