Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Hexyloxy)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A898135-1G A898135-5G A898135-100G A898135-10G A898135-250MG A898135-25G
103841-338EA 28 USD InStock
103841-338 76845-598 76845-600 77294-160 77294-162 77294-164 77294-166
4-(Hexyloxy)phenylboronic acid 98%
4-(Hexyloxy)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₉BO₃
Boiling Pt: 368.6 °C (760 mm Hg)
Melting Pt: 85 °C at N/A mmHg
Flash Pt: 176.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04039027
CAS Number: 121219-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty