Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,4,5-Tetracyanobenzene 97%

Supplier: AMBEED, INC
Warning

A829089-1G A829089-5G A829089-10G A829089-250MG A829089-25G
103841-162EA 97 USD InStock
103841-162 76845-396 76845-398 77199-842 77199-844 77199-846
1,2,4,5-Tetracyanobenzene 97%
1,2,4,5-Tetracyanobenzene
Precaución: for Research Use Only


Formula: C₁₀H₂N₄
Boiling Pt: 446.1 °C (760 mm Hg)
Melting Pt: 265…268 °C
Flash Pt: 235.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00043481
CAS Number: 712-74-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty