Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzil 98%

Supplier: AMBEED, INC
Warning

A563193-1000G A563193-100G A563193-500G A563193-25G
103810-454EA 35 USD InStock
103810-454 76696-230 76696-232 76843-994
Benzil 98%
Benzil
Precaución: for Research Use Only


Formula: (C₆H₅)C(O)C(O)(C₆H₅)
Boiling Pt: 347 °C (760 mm Hg)
Melting Pt: 94…97 °C
Flash Pt: 142.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00003080
CAS Number: 134-81-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty