Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[b]thiophene-3-boronic acid 98%

Supplier: AMBEED, INC
Warning

A229949-1G A229949-5G A229949-1G A229949-10G A229949-5G
103819-506EA 11 USD InStock
103819-506 103850-408 77089-264 77237-834 77303-860
Benzo[b]thiophene-3-boronic acid 98%
Benzo[b]thiophene-3-boronic acid
Precaución: for Research Use Only


Formula: C₈H₇BO₂S
Boiling Pt: 390.2 °C (760 mm Hg)
Melting Pt: 225…230 °C
Storage Temperature: Freezer
MDL Number: MFCD01114669
CAS Number: 113893-08-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty