Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,1,3-Benzoxadiazole-5-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A120934-1G A120934-100MG A120934-5G A120934-250MG
103800-010EA 104 USD InStock
103800-010 77136-488 77312-120 77685-060
2,1,3-Benzoxadiazole-5-carbaldehyde 97%
2,1,3-Benzoxadiazole-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Melting Pt: 56…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01200350
CAS Number: 32863-33-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty