Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[d][1,3]dioxol-4-amine 97%

Supplier: AMBEED, INC
Warning

A118425-1G A118425-100MG A118425-250MG A118425-1G A118425-5G
103799-052EA 53 USD InStock
103799-052 77189-500 77189-502 77297-598 77311-724
Benzo[d][1,3]dioxol-4-amine 97%
Benzo[d][1,3]dioxol-4-amine
Precaución: for Research Use Only


Formula: C₇H₇NO₂
Boiling Pt: 256.2 °C (760 mm Hg)
Flash Pt: 117.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038121
CAS Number: 1668-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty