Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Piperonylic acid 98%

Supplier: AMBEED, INC
Warning

A251901-5G A251901-100G A251901-10G A251901-500G A251901-25G A251901-5G
103851-854EA 5 USD InStock
103851-854 76874-632 76874-634 76874-636 77091-252 77304-568
Piperonylic acid 98%
Piperonylic acid
Precaución: for Research Use Only


Formula: C₈H₆O₄
MW: 166.13 g/mol
Boiling Pt: 180
Melting Pt: 229…231 °C
Flash Pt: 139.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00005830
CAS Number: 94-53-1
Merck Index: 12,07630
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty