Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[d]thiazol-5-ol 95%

Supplier: AMBEED, INC
Warning

A219408-1G A219408-5G A219408-250MG A219408-100MG
103818-792EA 308 USD InStock
103818-792 103849-668 77030-750 77872-112
Benzo[d]thiazol-5-ol 95%
Benzo[d]thiazol-5-ol
Precaución: for Research Use Only


Formula: C₇H₅NOS
Boiling Pt: 311.6 °C (760 mm Hg)
Flash Pt: 142.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00983978
CAS Number: 7686-41-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty