Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl (2-aminoethyl)carbamate 97%

Supplier: AMBEED, INC
Warning

A200050-1G A200050-5G A200050-10G A200050-5G A200050-1G A200050-25G
103817-160EA 10 USD InStock
103817-160 103847-786 76986-422 77085-844 77302-644 77302-646
Benzyl (2-aminoethyl)carbamate 97%
Benzyl (2-aminoethyl)carbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Melting Pt: 169…172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094510
CAS Number: 72080-83-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty