Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bicyclo[1.1.1]pentan-1-amine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A193519-1G A193519-5G A193519-250MG A193519-5G
103816-550EA 35 USD InStock
103816-550 103847-166 76873-130 77085-068
Bicyclo[1.1.1]pentan-1-amine hydrochloride 95%
Bicyclo[1.1.1]pentan-1-amine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₉N·HCl
MW: 119.59 g/mol
Melting Pt: 256…261 °C
Storage Temperature: Ambient
MDL Number: MFCD19204065
CAS Number: 22287-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty