Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bicyclo[4.2.0]octa-1,3,5-triene-3-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A384919-1G A384919-5G A384919-100MG A384919-250MG A384919-1G
103826-774EA 139 USD InStock
103826-774 103837-812 77251-946 77251-948 77306-966
Bicyclo[4.2.0]octa-1,3,5-triene-3-carboxylic acid 97%
Bicyclo[4.2.0]octa-1,3,5-triene-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₈O₂
MW: 148.15 g/mol
Boiling Pt: 306.1 °C (760 mm Hg)
Flash Pt: 137.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09842339
CAS Number: 875-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty