Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis((6-bromopyridin-2-yl)methyl)amine 95%

Supplier: AMBEED, INC
Warning

A102043-1G A102043-5G A102043-100MG A102043-250MG
103796-262EA 80 USD InStock
103796-262 103821-960 77160-238 77160-240
Bis((6-bromopyridin-2-yl)methyl)amine 95%
Bis((6-bromopyridin-2-yl)methyl)amine
Precaución: for Research Use Only


Formula: C₁₂H₁₁Br₂N₃
Boiling Pt: 434.5±40.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD28040561
CAS Number: 1265139-77-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty