Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2',4,4'-Tetrahydroxybenzophenone 98%

Supplier: AMBEED, INC
Warning

A193027-5G A193027-25G A193027-100G A193027-500G
103847-110EA 5 USD InStock
103847-110 77085-006 77248-600 77248-602
2,2',4,4'-Tetrahydroxybenzophenone 98%
2,2',4,4'-Tetrahydroxybenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₁₀O₅
MW: 246.22 g/mol
Melting Pt: 198…200 °C
Flash Pt: 289.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002278
CAS Number: 131-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty