Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2,6-dimethylphenyl)amine 97%

Supplier: AMBEED, INC
Warning

A138327-1G A138327-250MG A138327-1G A138327-5G
103802-598EA 92 USD InStock
103802-598 76823-342 77315-614 77315-616
Bis(2,6-dimethylphenyl)amine 97%
Bis(2,6-dimethylphenyl)amine
Precaución: for Research Use Only


Formula: C₁₆H₁₉N
Storage Temperature: Ambient
MDL Number: MFCD00168325
CAS Number: 74443-35-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty