Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-4-oxo-L-proline 97%

Supplier: AMBEED, INC
Warning

A135788-1G A135788-100G A135788-10G A135788-250MG A135788-1G A135788-25G A135788-500G
103802-218EA 8 USD InStock
103802-218 76996-630 76996-632 76996-634 77315-074 77315-076 77881-075
N-Boc-4-oxo-L-proline 97%
N-Boc-4-oxo-L-proline
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₅
MW: 229.23 g/mol
Melting Pt: 160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01860669
CAS Number: 84348-37-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty