Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Cha-OH 98%

Supplier: AMBEED, INC
Warning

A305715-1G A305715-100G A305715-10G A305715-1G A305715-25G A305715-5G
103814-446EA 9 USD InStock
103814-446 103835-612 76787-842 76787-844 76838-314 76838-316 76838-318
Boc-Cha-OH 98%
Boc-Cha-OH
Precaución: for Research Use Only


Formula: C₁₄H₂₅NO₄
Boiling Pt: 420.4 °C (760 mm Hg)
Flash Pt: 208.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00076896
CAS Number: 37736-82-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty