Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-d-homoserine 95%

Supplier: AMBEED, INC
Warning

A161455-1G A161455-5G A161455-1G A161455-10G A161455-250MG
103805-496EA 20 USD InStock
103805-496 103843-734 77319-718 77881-263 77881-264
Boc-d-homoserine 95%
Boc-d-homoserine
Precaución: for Research Use Only


Formula: C₉H₁₇NO₅
MW: 219.235 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12545888
CAS Number: 745011-75-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty