Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(tert-Butoxycarbonyl)guanidine 97%

Supplier: AMBEED, INC
Warning

A188466-1G A188466-5G A188466-10G A188466-25G A188466-1G A188466-5G
103816-104EA 5 USD InStock
103816-104 103846-700 76690-772 77084-444 77302-130 77302-132
1-(tert-Butoxycarbonyl)guanidine 97%
1-(tert-Butoxycarbonyl)guanidine
Precaución: for Research Use Only


Formula: C₆H₁₃N₃O₂
Boiling Pt: 259 °C (760 mm Hg)
Melting Pt: 170…175 °C
Flash Pt: 110.4 °C
Storage Temperature: Freezer
MDL Number: MFCD07369706
CAS Number: 219511-71-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty