Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-L-cyclopropylglycine 97%

Supplier: AMBEED, INC
Warning

A118794-1G A118794-100MG A118794-10G A118794-250MG A118794-1G A118794-5G
103799-108EA 18 USD InStock
103799-108 76688-016 76688-018 76688-020 77311-782 77311-784
Boc-L-cyclopropylglycine 97%
Boc-L-cyclopropylglycine
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₄
Boiling Pt: 364.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06659132
CAS Number: 155976-13-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty