Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-L-M-Tyrosine 98%

Supplier: AMBEED, INC
Warning

A109664-1G A109664-100MG A109664-250MG
103797-542EA 379 USD InStock
103797-542 77188-986 77188-988
Boc-L-M-Tyrosine 98%
Boc-L-M-Tyrosine
Precaución: for Research Use Only


Formula: C₁₄H₁₉NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD01632034
CAS Number: 90819-30-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty