Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-L-Pyroglutaminol 95%

Supplier: AMBEED, INC
Warning

A297556-1G A297556-5G A297556-1G A297556-250MG
103814-140EA 24 USD InStock
103814-140 103835-282 76837-968 76977-676
Boc-L-Pyroglutaminol 95%
Boc-L-Pyroglutaminol
Precaución: For research use only


Formula: C₁₀H₁₇NO₄
Boiling Pt: 370.9 °C (760 mm Hg)
Flash Pt: 178.09 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11111894
CAS Number: 81658-25-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty