Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(ɑ)-Boc-L-lysine 98%

Supplier: AMBEED, INC
Warning

A236606-1G A236606-5G A236606-100G A236606-10G A236606-500G A236606-1G A236606-25G A236606-5G
103811-112EA 6 USD InStock
103811-112 103850-886 76801-220 76801-222 76801-224 77089-942 77089-944 77089-946
N-(ɑ)-Boc-L-lysine 98%
N-(alpha)-Boc-L-lysine
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₄
MW: 246.3 g/mol
Boiling Pt: 410.5 °C (760 mm Hg)
Melting Pt: 205 °C
Flash Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00038203
CAS Number: 13734-28-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty