Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A130049-100G A130049-10G A130049-500G A130049-25G A130049-5G
77271-964EA 88 USD InStock
77271-964 77271-966 77271-968 77298-384 77298-386
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid
Precaución: for Research Use Only


Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Melting Pt: 58…60 °C
Flash Pt: 229.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00066063
CAS Number: 23680-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty