Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-L-tyrosine methyl ester 95%

Supplier: AMBEED, INC
Warning

A124268-1000G A124268-100G A124268-10G A124268-25G A124268-500G A124268-5G
76971-746EA 224 USD InStock
76971-746 76971-748 76971-750 76971-752 76971-754 77312-644
Boc-L-tyrosine methyl ester 95%
Boc-L-tyrosine methyl ester
Precaución: for Research Use Only


Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Boiling Pt: 452.7 °C (760 mm Hg)
Melting Pt: 100…104 °C
Flash Pt: 227.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00191181
CAS Number: 4326-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty