Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((4-(tert-Butyl)cyclohexyl)methyl)-3-hydroxynaphthalene-1,4-dione 98%

Supplier: AMBEED, INC
Warning

A101636-50MG A101636-100MG A101636-50MG A101636-1G A101636-250MG
103799-246EA 5 USD InStock
103799-246 76687-046 77282-778 77623-826 77876-784
2-((4-(tert-Butyl)cyclohexyl)methyl)-3-hydroxynaphthalene-1,4-dione 98%
2-((4-(tert-Butyl)cyclohexyl)methyl)-3-hydroxynaphthalene-1,4-dione
Precaución: for Research Use Only


Formula: C₂₁H₂₆O₃
Boiling Pt: 460.7 °C (760 mm Hg)
Melting Pt: 178…184 °C
Flash Pt: 246.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01712789
CAS Number: 88426-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty