Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3,4-Butanetetracarboxylic acid, (∼15% H₂O) 98% (contains < 15% H₂O)

Supplier: AMBEED, INC
Warning

A179548-10G A179548-1000G
103845-794EA 5 USD InStock
103845-794 76998-618 77881-359
1,2,3,4-Butanetetracarboxylic acid, (∼15% H₂O) 98% (contains < 15% H₂O)
1,2,3,4-Butanetetracarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₀O₈
Boiling Pt: 288.8 °C (760 mm Hg)
Melting Pt: 195…197 °C
Flash Pt: 142.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00002722
CAS Number: 1703-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty