Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cariporide 97%

Supplier: AMBEED, INC
Warning

A217757-5MG A217757-10MG A217757-50MG A217757-1MG A217757-10MG A217757-50MG A217757-5MG A217757-100MG
103791-564EA 73 USD InStock
103791-564 103792-428 103799-496 76691-678 77088-014 77088-016 77088-018 77877-680
Cariporide 97%
Cariporide
Precaución: for Research Use Only


Formula: C₁₂H₁₇N₃O₃S
MW: 283.34 g/mol
Boiling Pt: 542.8 °C (760 mm Hg)
Flash Pt: 282.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00864683
CAS Number: 159138-80-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty