Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carnosol 95%

Supplier: AMBEED, INC
Warning

A833913-1MG A833913-100MG A833913-1G A833913-250MG
103792-856EA 52 USD InStock
103792-856 103792-858 76698-272 77627-638 77878-684 77878-685
Carnosol 95%
Carnosol
Precaución: for Research Use Only


Formula: C₂₀H₂₆O₄
Boiling Pt: 524.8 °C (760 mm Hg)
Flash Pt: 187 °C
Storage Temperature: Freezer
MDL Number: MFCD02752467
CAS Number: 5957-80-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty