Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorphenoxamine ≥98%

Supplier: AMBEED, INC
Warning

A629750-50MG A629750-100MG A629750-1G A629750-250MG
103792-792EA 25 USD InStock
103792-792 103799-702 77627-260 77880-336 77880-337
Chlorphenoxamine ≥98%
Chlorphenoxamine
Precaución: Sealed in dry, at room temperature.


Formula: C₁₈H₂₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD00866588
CAS Number: 77-38-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty