Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chroman-4-amine 97%

Supplier: AMBEED, INC
Warning

A108900-1G A108900-250MG A108900-1G A108900-5G
103797-400EA 160 USD InStock
103797-400 103823-190 77160-616 77310-020 77310-022
Chroman-4-amine 97%
Chroman-4-amine
Precaución: for Research Use Only


Formula: C₉H₁₁NO
Boiling Pt: 237.4 °C (760 mm Hg)
Flash Pt: 102.106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00730969
CAS Number: 53981-38-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty