Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-3-Aminocyclobutanol hydrochloride 97%

Supplier: AMBEED, INC
Warning

A197398-5G A197398-100MG A197398-250MG A197398-1G A197398-5G
103816-918EA 24 USD InStock
103816-918 103847-540 76814-132 76814-134 77085-548 77085-550
cis-3-Aminocyclobutanol hydrochloride 97%
cis-3-Aminocyclobutanol hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₀ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD17214231
CAS Number: 1219019-22-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty