Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Hexahydroisoindole hydrochloride 95%

Supplier: AMBEED, INC
Warning

A143684-1G A143684-100G A143684-1G A143684-25G A143684-5G
103803-310EA 8 USD InStock
103803-310 76811-866 77316-604 77316-606 77316-608
cis-Hexahydroisoindole hydrochloride 95%
cis-Hexahydroisoindole hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₅N·ClH
Storage Temperature: Ambient
MDL Number: MFCD09026400
CAS Number: 161829-92-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty