Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Bicyclo[3.3.0]octane-3,7-dione 97%

Supplier: AMBEED, INC
Warning

A132031-1G A132031-100G A132031-250MG A132031-25G A132031-1G A132031-5G
103801-678EA 11 USD InStock
103801-678 76688-666 76688-668 76688-670 77314-224 77314-226
cis-Bicyclo[3.3.0]octane-3,7-dione 97%
cis-Bicyclo[3.3.0]octane-3,7-dione
Precaución: for Research Use Only


Formula: C₈H₁₀O₂
Boiling Pt: 268.3 °C (760 mm Hg)
Melting Pt: 83…86 °C
Flash Pt: 99.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00003771
CAS Number: 51716-63-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty