Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cloxiquina 98%

Supplier: AMBEED, INC
Warning

A162839-1000G A162839-100G A162839-500G A162839-25G
103843-920EA 5 USD InStock
103843-920 76997-880 76997-882 76997-884 76997-886 77319-994
Cloxiquina 98%
Cloxiquina
Precaución: for Research Use Only


Formula: C₉H₆ClNO
Melting Pt: 122…124 °C
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00006788
CAS Number: 130-16-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty