Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cycloheptane-1,3-dione 97%

Supplier: AMBEED, INC
Warning

A156409-1G A156409-5G A156409-10G A156409-250MG A156409-1G A156409-25G A156409-5G
103804-874EA 19 USD InStock
103804-874 103843-074 77190-958 77190-960 77300-178 77300-180 77318-848
Cycloheptane-1,3-dione 97%
Cycloheptane-1,3-dione
Precaución: Inert atmosphere, room temperature


Formula: C₇H₁₀O₂
Boiling Pt: 250.7 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01863735
CAS Number: 1194-18-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty